Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyg11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033634.3 | NP_001028806.2 | 3 - 4 | 1122 | 83 | |
GenBank mRNA | AK145130.1 | - | 3 - 4 | 1095 | 83 | |
AK148335.1 | - | 3 - 4 | 1121 | 83 | ||
AK162646.1 | - | 3 - 4 | 1085 | 83 | ||
AK166031.1 | - | 3 - 4 | 1102 | 83 | ||
AK166174.1 | - | 3 - 4 | 1102 | 83 | ||
AK166278.1 | - | 3 - 4 | 1112 | 83 | ||
BC117977.2 | - | 3 - 4 | 1086 | 40099428 | 83 | |
BC117978.2 | - | 3 - 4 | 1086 | 40099429 | 83 | |
BC139383.1 | - | 3 - 4 | 1089 | 8862405 | 83 | |
BC139384.1 | - | 3 - 4 | 1089 | 8862406 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |