Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027126.4 | NP_081402.3 | 1 - 2 | 145 | 79 | |
GenBank mRNA | AJ557515.1 | - | 1 - 2 | 145 | 79 | |
AK014082.1 | - | 1 - 2 | 117 | 79 | ||
BC022603.1 | - | 1 - 2 | 169 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |