Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172694.2 | NP_766282.1 | 4 - 5 | 1182 | 66 | |
GenBank mRNA | AK035987.1 | - | 4 - 5 | 1183 | 66 | |
AK039319.1 | - | 4 - 5 | 1183 | 66 | ||
AK122378.1 | - | 1173 | 66 | |||
BC088994.1 | - | 4 - 5 | 1091 | 6734479 | 66 | |
BC138128.1 | - | 4 - 5 | 1121 | 8861147 | 66 | |
BC138129.1 | - | 4 - 5 | 1121 | 8861148 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |