Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007400.2 | NP_031426.2 | 11 - 12 | 1370 | 65 | |
GenBank mRNA | AK051780.1 | - | 1 - 2 | 184 | 65 | |
D50411.1 | - | 11 - 12 | 1370 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |