Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172748.2 | NP_766336.2 | 2 - 3 | 565 | 118 | |
GenBank mRNA | AK028405.1 | - | 2 - 3 | 334 | 118 | |
AK031431.1 | - | 1 - 2 | 306 | 118 | ||
BC059812.1 | - | 2 - 3 | 565 | 6401846 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |