Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53i11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025246.1 | NP_001020417.1 | 1 - 2 | 170 | 134 | |
GenBank mRNA | BC098206.1 | - | 1 - 2 | 170 | 6812412 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |