Search Thermo Fisher Scientific
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010806.1 | NP_034936.1 | 13 - 14 | 1899 | 64 | |
GenBank mRNA | AK016557.1 | - | 12 - 13 | 2018 | 64 | |
CK637291.1 | - | 4 - 5 | 551 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |