Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030138.1 | - | 3 - 4 | 283 | 107 | |
AK030182.1 | - | 3 - 4 | 309 | 107 | ||
AK040259.1 | - | 3 - 4 | 198 | 107 | ||
AK079691.1 | - | 3 - 4 | 199 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |