Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025302.3 | NP_079578.1 | 5 - 6 | 691 | 96 | |
GenBank mRNA | AK007672.1 | - | 5 - 6 | 679 | 96 | |
AK009521.1 | - | 5 - 6 | 685 | 96 | ||
BC027762.1 | - | 5 - 6 | 692 | 5353459 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |