Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175113.3 | NP_780322.2 | 3 - 4 | 485 | 125 | |
GenBank mRNA | AK014359.1 | - | 3 - 4 | 482 | 125 | |
AK017672.1 | - | 3 - 4 | 427 | 125 | ||
AK028755.1 | - | 3 - 4 | 485 | 125 | ||
AK129300.1 | - | 3 - 4 | 452 | 125 | ||
AK164396.1 | - | 3 - 4 | 418 | 125 | ||
AK165976.1 | - | 3 - 4 | 437 | 125 | ||
AK167222.1 | - | 3 - 4 | 451 | 125 | ||
BC027565.1 | - | 3 - 4 | 391 | 125 | ||
BC052648.1 | - | 3 - 4 | 403 | 30030801 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |