Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178385.3 | NP_848472.2 | 1 | 1710 | 71 | |
GenBank mRNA | AK132036.1 | - | 1660 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |