Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025867.2 | NP_080143.1 | 6 - 7 | 674 | 152 | |
GenBank mRNA | AK009003.1 | - | 6 - 7 | 676 | 152 | |
AK009855.1 | - | 6 - 7 | 675 | 152 | ||
AY367772.1 | - | 5 - 6 | 610 | 152 | ||
BC010313.1 | - | 5 - 6 | 2646766 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |