Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175686.3 | NP_783617.2 | 3 - 4 | 1612 | 112 | |
GenBank mRNA | AK008428.2 | - | 4 - 5 | 442 | 112 | |
AK019503.1 | - | 3 - 4 | 423 | 112 | ||
BC092372.1 | - | 3 - 4 | 961 | 30533016 | 112 | |
L06502.1 | - | 3 - 4 | 777 | 112 | ||
U03873.1 | - | 3 - 4 | 1618 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |