Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029004.1 | NP_083280.1 | 2 - 3 | 319 | 56 | |
GenBank mRNA | AK018593.1 | - | 2 - 3 | 308 | 56 | |
AK039330.1 | - | 683 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |