Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291059.1 | NP_001277988.1 | 8 - 9 | 1249 | 127 | |
NM_001291062.1 | NP_001277991.1 | 5 - 6 | 844 | 127 | ||
NM_001291063.1 | NP_001277992.1 | 5 - 6 | 880 | 127 | ||
NM_023502.2 | NP_075991.1 | 8 - 9 | 1285 | 127 | ||
GenBank mRNA | AF256216.1 | - | 7 - 8 | 970 | 127 | |
AF256217.1 | - | 7 - 8 | 1006 | 127 | ||
AF256218.1 | - | 5 - 6 | 724 | 127 | ||
AF256219.1 | - | 5 - 6 | 760 | 127 | ||
AK088422.1 | - | 5 - 6 | 845 | 127 | ||
AK134180.1 | - | 3 - 4 | 315 | 127 | ||
AK146819.1 | - | 8 - 9 | 1250 | 127 | ||
AK160118.1 | - | 8 - 9 | 1155 | 127 | ||
BC051937.1 | - | 7 - 8 | 1056 | 127 | ||
BC092057.1 | - | 7 - 8 | 1028 | 4016339 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |