Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201357.2 | NP_958745.2 | 4 - 5 | 547 | 96 | |
GenBank mRNA | AK150308.1 | - | 4 - 5 | 548 | 96 | |
AK151830.1 | - | 4 - 5 | 548 | 96 | ||
BC031458.1 | - | 4 - 5 | 510 | 4191954 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |