Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286002.1 | NP_001272931.1 | 6 - 7 | 912 | 58 | |
NM_028776.5 | NP_083052.1 | 6 - 7 | 1099 | 58 | ||
GenBank mRNA | AK004809.2 | - | 6 - 7 | 839 | 58 | |
AK030824.1 | - | 6 - 7 | 1074 | 58 | ||
AK162937.1 | - | 6 - 7 | 913 | 58 | ||
BC043085.1 | - | 6 - 7 | 800 | 6417440 | 58 | |
BC066800.1 | - | 6 - 7 | 784 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |