Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181405.4 | NP_852070.3 | 4 - 5 | 1064 | 63 | |
GenBank mRNA | AK154881.1 | - | 4 - 5 | 1065 | 63 | |
BC031789.1 | - | 4 - 5 | 484 | 4006916 | 63 | |
BQ559332.1 | - | 2 - 3 | 248 | 63 | ||
CA315475.1 | - | 4 - 5 | 758 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |