Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013381.2 | NP_001013399.1 | 1 - 2 | 226 | 54 | |
GenBank mRNA | AK148401.1 | - | 1 - 2 | 221 | 54 | |
BC139274.1 | - | 1 - 2 | 169 | 8862296 | 54 | |
BC139276.1 | - | 1 - 2 | 169 | 8862298 | 54 | |
BC145594.1 | - | 1 - 2 | 169 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |