Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025825.3 | NP_080101.1 | 4 - 5 | 810 | 145 | |
GenBank mRNA | AK005440.1 | - | 4 - 5 | 516 | 145 | |
AK122222.1 | - | 417 | 145 | |||
AK133853.1 | - | 4 - 5 | 802 | 145 | ||
AK207768.1 | - | 1 - 2 | 119 | 145 | ||
AK217461.1 | - | 1 - 2 | 83 | 145 | ||
BC018442.1 | - | 4 - 5 | 788 | 3157290 | 145 | |
BC019152.1 | - | 4 - 5 | 811 | 5065139 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |