Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029447.2 | NP_083723.1 | 9 - 10 | 1687 | 87 | |
GenBank mRNA | AK079090.1 | - | 9 - 10 | 1688 | 87 | |
AK138399.1 | - | 9 - 10 | 1660 | 87 | ||
AK146364.1 | - | 9 - 10 | 1636 | 87 | ||
AK170508.1 | - | 9 - 10 | 1688 | 87 | ||
AK171432.1 | - | 9 - 10 | 1688 | 87 | ||
BC016224.1 | - | 9 - 10 | 1660 | 4486941 | 87 | |
BC025520.1 | - | 2 - 3 | 546 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |