Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026396.3 | NP_080672.3 | 6 - 7 | 549 | 98 | |
GenBank mRNA | AK002985.1 | - | 6 - 7 | 436 | 98 | |
AK168325.1 | - | 6 - 7 | 522 | 98 | ||
BC014832.1 | - | 6 - 7 | 519 | 4484024 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |