Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF126967.1 | - | 1 - 2 | 176 | 56 | |
AK033285.1 | - | 1 - 2 | 185 | 56 | ||
BC005634.1 | - | 1 - 2 | 105 | 56 | ||
BC031757.1 | - | 1 - 2 | 159 | 56 | ||
BC046459.1 | - | 1 - 2 | 243 | 5322519 | 56 | |
BC071228.1 | - | 1 - 2 | 188 | 4915902 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |