Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053249.3 | NP_444479.2 | 5 - 6 | 991 | 73 | |
GenBank mRNA | AY028606.1 | - | 5 - 6 | 999 | 73 | |
BC108971.2 | - | 5 - 6 | 991 | 40047321 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |