Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163691.1 | NP_001157163.1 | 32 - 33 | 6038 | 116 | |
NM_021415.4 | NP_067390.4 | 32 - 33 | 6038 | 116 | ||
GenBank mRNA | AF051947.1 | - | 1 - 2 | 259 | 116 | |
AK122452.1 | - | 21 - 22 | 3011 | 116 | ||
AK135951.1 | - | 14 - 15 | 1785 | 116 | ||
AK147242.1 | - | 32 - 33 | 6039 | 116 | ||
BC138026.1 | - | 32 - 33 | 5877 | 8861045 | 116 | |
CF744603.1 | - | 1 - 2 | 110 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |