Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026214.2 | NP_001021385.1 | 12 - 13 | 1154 | 88 | |
NM_001286049.1 | NP_001272978.1 | 11 - 12 | 1129 | 88 | ||
NM_001286058.1 | NP_001272987.1 | 10 - 11 | 1056 | 88 | ||
NM_007647.3 | NP_031673.2 | 11 - 12 | 1081 | 88 | ||
GenBank mRNA | AF006482.1 | - | 11 - 12 | 1089 | 88 | |
AF136571.1 | - | 11 - 12 | 1115 | 88 | ||
AJ238636.1 | - | 11 - 12 | 1117 | 88 | ||
AK002618.1 | - | 11 - 12 | 1054 | 88 | ||
AK031581.1 | - | 12 - 13 | 1134 | 88 | ||
AK036641.1 | - | 12 - 13 | 1131 | 88 | ||
AK037736.1 | - | 11 - 12 | 1050 | 88 | ||
AK045828.1 | - | 11 - 12 | 1067 | 88 | ||
AK079267.1 | - | 10 - 11 | 978 | 88 | ||
AK080265.1 | - | 12 - 13 | 1141 | 88 | ||
AK081435.1 | - | 12 - 13 | 1119 | 88 | ||
AK088455.1 | - | 11 - 12 | 1110 | 88 | ||
AK160950.1 | - | 9 - 10 | 975 | 88 | ||
BC015247.1 | - | 10 - 11 | 1039 | 4242560 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |