Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195003.1 | NP_001181932.1 | 7 - 8 | 1045 | 66 | |
NM_001195004.1 | NP_001181933.1 | 6 - 7 | 955 | 66 | ||
NM_177619.3 | NP_808287.1 | 5 - 6 | 778 | 66 | ||
GenBank mRNA | AK147968.1 | - | 6 - 7 | 772 | 66 | |
AK170389.1 | - | 6 - 7 | 902 | 66 | ||
AK186978.1 | - | 2 - 3 | 130 | 66 | ||
BC024092.1 | - | 5 - 6 | 618 | 66 | ||
BC048904.1 | - | 5 - 6 | 733 | 6492124 | 66 | |
BC057102.1 | - | 6 - 7 | 883 | 6401353 | 66 | |
DQ076247.1 | - | 7 - 8 | 973 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |