Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030225.4 | NP_084501.1 | 8 - 9 | 661 | 119 | |
GenBank mRNA | AK019713.2 | - | 658 | 119 | ||
AK054053.1 | - | 8 - 9 | 615 | 119 | ||
AK149664.1 | - | 8 - 9 | 637 | 119 | ||
AK158877.1 | - | 8 - 9 | 662 | 119 | ||
AK168570.1 | - | 8 - 9 | 637 | 119 | ||
AK169943.1 | - | 8 - 9 | 615 | 119 | ||
AK194738.1 | - | 1 - 2 | 67 | 119 | ||
BC006702.1 | - | 8 - 9 | 598 | 3583936 | 119 | |
CT010197.1 | - | 8 - 9 | 597 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |