Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172853.2 | NP_766441.1 | 7 - 8 | 1584 | 68 | |
GenBank mRNA | AK034096.1 | - | 7 - 8 | 1584 | 68 | |
AK137369.1 | - | 7 - 8 | 1507 | 68 | ||
AY267034.1 | - | 6 - 7 | 1439 | 68 | ||
BC083189.1 | - | 8 - 9 | 2174 | 68 | ||
BC141119.1 | - | 6 - 7 | 1415 | 9055731 | 68 | |
BC145423.1 | - | 7 - 8 | 1533 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |