Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164593.1 | NP_001158065.1 | 2 - 3 | 780 | 96 | |
GenBank mRNA | BC151034.1 | - | 2 - 3 | 709 | 9087947 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |