Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033633.3 | NP_001028805.2 | 4 - 5 | 1169 | 65 | |
GenBank mRNA | AK147341.1 | - | 4 - 5 | 1170 | 65 | |
BC119586.1 | - | 5 - 6 | 736 | 40058865 | 65 | |
BC119587.1 | - | 4 - 5 | 678 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |