Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026775.4 | NP_081051.1 | 2 - 3 | 394 | 82 | |
GenBank mRNA | AK003676.1 | - | 2 - 3 | 377 | 82 | |
AK084117.1 | - | 2 - 3 | 377 | 82 | ||
AK150605.1 | - | 2 - 3 | 366 | 82 | ||
AK152643.1 | - | 2 - 3 | 377 | 82 | ||
AK152656.1 | - | 2 - 3 | 366 | 82 | ||
AK186798.1 | - | 2 - 3 | 139 | 82 | ||
BC064755.1 | - | 2 - 3 | 368 | 30029180 | 82 | |
BC085097.1 | - | 2 - 3 | 373 | 5349826 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |