Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200005027.mm.1 |
NM_019579.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005027.mm.1 |
NM_019579.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005027.mm.1 |
TC1200000711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005029.mm.1 |
NM_019579.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005029.mm.1 |
NM_019579.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005029.mm.1 |
TC1200000711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005026.mm.1 |
NM_019579.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005026.mm.1 |
NM_019579.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005026.mm.1 |
TC1200000711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005028.mm.1 |
NM_019579.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005028.mm.1 |
NM_019579.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005028.mm.1 |
TC1200000711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005031.mm.1 |
NM_019579.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005031.mm.1 |
NM_019579.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005031.mm.1 |
TC1200000711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200005030.mm.1 |
NM_019579.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005030.mm.1 |
NM_019579.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200005030.mm.1 |
TC1200000711.mm.1 |
MTA TC |