Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037914.3 | NP_001033003.1 | 1 - 2 | 120 | 96 | |
GenBank mRNA | AK134107.1 | - | 1 - 2 | 121 | 96 | |
BC150962.1 | - | 1 - 2 | 143 | 9087873 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |