Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029427.1 | NP_083703.1 | 1 - 2 | 256 | 68 | |
GenBank mRNA | AK014771.1 | - | 1 - 2 | 257 | 68 | |
BC109360.2 | - | 1 - 2 | 256 | 40058461 | 68 | |
BC109361.2 | - | 1 - 2 | 256 | 40058463 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |