Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080440.3 | NP_536688.2 | 2 - 3 | 2387 | 140 | |
GenBank mRNA | AF453257.1 | - | 2 - 3 | 2388 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |