Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114334.1 | NP_001107806.1 | 5 - 6 | 634 | 114 | |
GenBank mRNA | AK032724.1 | - | 5 - 6 | 596 | 114 | |
AK135716.1 | - | 5 - 6 | 602 | 114 | ||
AK163258.1 | - | 5 - 6 | 604 | 114 | ||
BC038491.1 | - | 5 - 6 | 607 | 4010710 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |