Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008246.2 | NP_032272.2 | 1 - 2 | 281 | 93 | |
GenBank mRNA | AK004779.1 | - | 1 - 2 | 220 | 93 | |
AK135865.1 | - | 1 - 2 | 102 | 93 | ||
AK158838.1 | - | 1 - 2 | 282 | 93 | ||
AK169626.1 | - | 1 - 2 | 221 | 93 | ||
BC118047.1 | - | 1 - 2 | 101 | 40106779 | 93 | |
D88315.1 | - | 1 - 2 | 154 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |