Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159672.1 | NP_001153144.1 | 2 - 3 | 139 | 111 | |
NM_175317.3 | NP_780526.2 | 2 - 3 | 168 | 111 | ||
GenBank mRNA | AK028676.1 | - | 2 - 3 | 141 | 111 | |
AK031647.1 | - | 2 - 3 | 125 | 111 | ||
AK049698.1 | - | 2 - 3 | 126 | 111 | ||
BC045616.1 | - | 2 - 3 | 122 | 4983234 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |