Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172310.2 | NP_758514.2 | 15 - 16 | 1980 | 71 | |
GenBank mRNA | AK040927.1 | - | 15 - 16 | 1953 | 71 | |
AK041280.1 | - | 15 - 16 | 1980 | 71 | ||
AK079416.1 | - | 15 - 16 | 1962 | 71 | ||
BC039225.1 | - | 15 - 16 | 1964 | 4458085 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |