Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 19 - 20 | 2965 | 129 | |
GenBank mRNA | AF093576.1 | - | 19 - 20 | 2873 | 129 | |
BC150747.1 | - | 19 - 20 | 2830 | 9087658 | 129 | |
U87455.1 | - | 19 - 20 | 2945 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |