Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008689.2 | NP_032715.2 | 19 - 20 | 2607 | 91 | |
GenBank mRNA | AB119195.1 | - | 17 - 18 | 2105 | 91 | |
AB221614.1 | - | 17 - 18 | 2112 | 91 | ||
AK089660.1 | - | 7 - 8 | 981 | 91 | ||
AK146389.1 | - | 5 - 6 | 826 | 91 | ||
AK157915.1 | - | 4 - 5 | 607 | 91 | ||
AK159423.1 | - | 7 - 8 | 1038 | 91 | ||
AY521462.1 | - | 17 - 18 | 2105 | 91 | ||
AY521463.1 | - | 17 - 18 | 2105 | 91 | ||
BC138535.1 | - | 18 - 19 | 2184 | 8861555 | 91 | |
BC138536.1 | - | 18 - 19 | 2184 | 8861556 | 91 | |
BC145317.1 | - | 19 - 20 | 2201 | 91 | ||
L28117.1 | - | 7 - 8 | 90 | |||
M57999.1 | - | 19 - 20 | 2396 | 91 | ||
S66656.1 | - | 7 - 8 | 1151 | 91 | ||
S89033.1 | - | 7 - 8 | 1199 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |