Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145921.1 | NP_001139393.1 | 4 - 5 | 1758 | 60 | |
NM_001145922.1 | NP_001139394.1 | 4 - 5 | 1758 | 60 | ||
NM_198249.4 | NP_937892.2 | 4 - 5 | 1758 | 60 | ||
GenBank mRNA | AK131136.1 | - | 4 - 5 | 1727 | 60 | |
AK143527.1 | - | 4 - 5 | 1733 | 60 | ||
AK147761.1 | - | 4 - 5 | 1694 | 60 | ||
AK155659.1 | - | 3 - 4 | 2777 | 60 | ||
AK163945.1 | - | 4 - 5 | 1759 | 60 | ||
BC059018.1 | - | 4 - 5 | 1660 | 6848691 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |