Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026474.5 | NP_080750.1 | 6 - 7 | 515 | 125 | |
GenBank mRNA | AK007881.1 | - | 6 - 7 | 450 | 125 | |
AK010711.1 | - | 5 - 6 | 484 | 125 | ||
AK010830.1 | - | 6 - 7 | 432 | 125 | ||
AK020433.1 | - | 6 - 7 | 434 | 125 | ||
AK045153.1 | - | 6 - 7 | 454 | 125 | ||
AK082328.1 | - | 6 - 7 | 517 | 125 | ||
AK141416.1 | - | 451 | 125 | |||
AK148838.1 | - | 6 - 7 | 429 | 125 | ||
BC009167.1 | - | 6 - 7 | 432 | 2811924 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |