Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008608.3 | NP_032634.3 | 7 - 8 | 1413 | 96 | |
GenBank mRNA | AK040968.1 | - | 1386 | 96 | ||
AK088476.1 | - | 7 - 8 | 1413 | 96 | ||
AK138611.1 | - | 7 - 8 | 1354 | 96 | ||
AK149636.1 | - | 7 - 8 | 1342 | 96 | ||
AK149907.1 | - | 7 - 8 | 1393 | 96 | ||
AK149988.1 | - | 7 - 8 | 1313 | 96 | ||
AK155117.1 | - | 7 - 8 | 1415 | 96 | ||
AK163601.1 | - | 7 - 8 | 1405 | 96 | ||
AK165014.1 | - | 7 - 8 | 1414 | 96 | ||
AK212086.1 | - | 3 - 4 | 364 | 96 | ||
BC076638.1 | - | 7 - 8 | 1405 | 30658833 | 96 | |
DQ249870.1 | - | 7 - 8 | 1151 | 96 | ||
U54984.1 | - | 7 - 8 | 1397 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |