Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033713.2 | NP_001028885.1 | 4 - 5 | 587 | 70 | |
NM_001291195.1 | NP_001278124.1 | 4 - 5 | 587 | 70 | ||
GenBank mRNA | AK132678.1 | - | 4 - 5 | 469 | 70 | |
BC019116.1 | - | 4 - 5 | 573 | 70 | ||
BC061128.1 | - | 4 - 5 | 558 | 6593862 | 70 | |
U30823.1 | - | 2 - 3 | 336 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |