Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdadc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168535.1 | NP_001162007.1 | 7 - 8 | 1342 | 112 | |
NM_001168536.1 | NP_001162008.1 | 7 - 8 | 1342 | 112 | ||
NM_001168537.1 | NP_001162009.1 | 7 - 8 | 1342 | 112 | ||
NM_001168538.1 | NP_001162010.1 | 7 - 8 | 1342 | 112 | ||
NM_027986.3 | NP_082262.1 | 7 - 8 | 1342 | 112 | ||
GenBank mRNA | AK009280.1 | - | 7 - 8 | 1334 | 112 | |
AK032600.1 | - | 7 - 8 | 1321 | 112 | ||
AK032813.1 | - | 7 - 8 | 1328 | 112 | ||
AK038926.1 | - | 7 - 8 | 1322 | 112 | ||
AK039148.1 | - | 7 - 8 | 1330 | 112 | ||
BC006901.1 | - | 7 - 8 | 1304 | 3600168 | 112 | |
BE648840.1 | - | 1 - 2 | 89 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |