Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027808.1 | NP_082084.1 | 10 - 11 | 3471 | 92 | |
GenBank mRNA | AK173191.1 | - | 9 - 10 | 5183 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |