Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177992.2 | NP_818773.1 | 3 - 4 | 359 | 67 | |
GenBank mRNA | AK031004.1 | - | 3 - 4 | 361 | 67 | |
AK075772.1 | - | 3 - 4 | 317 | 67 | ||
BC003886.1 | - | 3 - 4 | 318 | 3590411 | 67 | |
BC024109.1 | - | 3 - 4 | 315 | 5149353 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |