Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289729.1 | NP_001276658.1 | 3 - 4 | 651 | 77 | |
NM_133213.2 | NP_573476.2 | 3 - 4 | 651 | 77 | ||
GenBank mRNA | AF367247.1 | - | 3 - 4 | 315 | 77 | |
AK078835.1 | - | 3 - 4 | 458 | 77 | ||
AK085520.1 | - | 3 - 4 | 449 | 77 | ||
AK087627.1 | - | 3 - 4 | 458 | 77 | ||
AK164538.1 | - | 3 - 4 | 479 | 77 | ||
AK172086.1 | - | 3 - 4 | 652 | 77 | ||
BC140977.1 | - | 3 - 4 | 295 | 9055589 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |